siRNA Details
| Gene Symbol: Trh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013046.3 | NP_037178.1 | 2, 3 | 344 | ||
| XM_006236899.2 | XP_006236961.1 | 2, 3 | 775 | |||
| GenBank mRNA | BC161830.1 | - | 2, 3 | 291 | 9091055 | |
| FQ100942.1 | - | 2, 3 | 344 | |||
| FQ221570.1 | - | 2, 3 | 320 | |||
| FQ221573.1 | - | 2, 3 | 320 | |||
| FQ222015.1 | - | 2, 3 | 320 | |||
| FQ222264.1 | - | 2, 3 | 318 | |||
| FQ222377.1 | - | 2, 3 | 319 | |||
| FQ222571.1 | - | 2, 3 | 318 | |||
| FQ222984.1 | - | 2, 3 | 318 | |||
| FQ223091.1 | - | 2, 3 | 320 | |||
| FQ228499.1 | - | 2, 3 | 320 | |||
| FQ228758.1 | - | 2, 3 | 320 | |||
| FQ228762.1 | - | 2, 3 | 318 | |||
| FQ229082.1 | - | 2, 3 | 320 | |||
| FQ229292.1 | - | 2, 3 | 318 | |||
| M12138.1 | - | 2, 3 | 311 | |||
| M36317.1 | - | 2, 3 | 311 | |||

Trh
Genomic Map