siRNA Details
| Gene Symbol: Csf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017104.1 | NP_058800.1 | 2 | 149 | ||
| XM_006247265.2 | XP_006247327.2 | 2 | 218 | |||
| GenBank mRNA | U37101.1 | - | 2 | 149 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Csf3| Gene Symbol: Csf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017104.1 | NP_058800.1 | 2 | 149 | ||
| XM_006247265.2 | XP_006247327.2 | 2 | 218 | |||
| GenBank mRNA | U37101.1 | - | 2 | 149 | ||