siRNA Details
| Gene Symbol: Il7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013110.2 | NP_037242.2 | 3 | 178 | ||
| XM_006232139.2 | XP_006232201.1 | 3 | 3024 | |||
| XM_008760860.1 | XP_008759082.1 | 3 | 3024 | |||
| GenBank mRNA | AF010464.1 | - | 3 | 187 | ||
| AF367210.1 | - | 3 | 178 | |||
| FQ226544.1 | - | 748 | ||||
| FQ226549.1 | - | 748 | ||||

Il7
Genomic Map