siRNA Details
| Gene Symbol: Tshb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013116.2 | NP_037248.2 | 2, 3 | 224 | ||
| XM_008761373.1 | XP_008759595.1 | 2, 3 | 2355 | |||
| GenBank mRNA | BC058488.1 | - | 2, 3 | 184 | 6921292 | |
| CB315383.1 | - | 2, 3 | 183 | |||
| D00578.1 | - | 1, 2 | 37 | |||
| M10902.1 | - | 2, 3 | 179 | |||
| X01454.1 | - | 2, 3 | 222 | |||

Tshb
Genomic Map