siRNA Details
| Gene Symbol: Tshb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013116.2 | NP_037248.2 | 3 | 309 | ||
| XM_008761373.1 | XP_008759595.1 | 3 | 2440 | |||
| GenBank mRNA | BC058488.1 | - | 3 | 269 | 6921292 | |
| CB315383.1 | - | 3 | 268 | |||
| D00578.1 | - | 2 | 122 | |||
| M10902.1 | - | 3 | 264 | |||
| X01454.1 | - | 3 | 307 | |||

Tshb
Genomic Map