siRNA Details
Gene Symbol: Ppard | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013141.2 | NP_037273.2 | 5 | 564 | ||
XM_006256166.2 | XP_006256228.1 | 5 | 1376 | |||
XM_006256167.2 | XP_006256229.1 | 4 | 1483 | |||
XM_006256168.2 | XP_006256230.1 | 4 | 2378 | |||
XM_008772797.1 | XP_008771019.1 | 4 | 1611 | |||
GenBank mRNA | AJ306400.1 | - | 3 | 330 | ||
FM033221.1 | - | 5 | 564 | |||
U40064.1 | - | 5 | 585 | |||
U75918.1 | - | 5 | 578 |
