siRNA Details
Gene Symbol: Mxi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013160.2 | NP_037292.2 | 5 | 516 | ||
XM_006231604.1 | XP_006231666.1 | 5 | 1075 | |||
XM_006231605.1 | XP_006231667.1 | 5 | 487 | |||
XM_006231606.1 | XP_006231668.1 | 5 | 646 | |||
GenBank mRNA | BC081824.1 | - | 5 | 516 | 7133853 |
