siRNA Details
| Gene Symbol: Ifng | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138880.2 | NP_620235.1 | 3 | 356 | ||
| GenBank mRNA | AF010466.1 | - | 3 | 356 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ifng| Gene Symbol: Ifng | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138880.2 | NP_620235.1 | 3 | 356 | ||
| GenBank mRNA | AF010466.1 | - | 3 | 356 | ||