siRNA Details
| Gene Symbol: Aptx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_148889.2 | NP_683687.1 | 4 | 487 | ||
| XM_006238026.2 | XP_006238088.1 | 4 | 843 | |||
| XM_006238027.2 | XP_006238089.1 | 3 | 879 | |||
| XM_006238028.2 | XP_006238090.1 | 3 | 726 | |||
| XM_006238029.2 | XP_006238091.1 | 2 | 60 | |||
| XM_008763607.1 | XP_008761829.1 | 2 | 738 | |||
| GenBank mRNA | AF398235.1 | - | 3 | 233 | ||
| BC078716.1 | - | 4 | 487 | 7132044 | ||

Aptx
Genomic Map