siRNA Details
| Gene Symbol: Spink1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152936.1 | NP_690919.1 | 2, 3 | 159 | ||
| GenBank mRNA | M27883.1 | - | 2, 3 | 159 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spink1| Gene Symbol: Spink1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152936.1 | NP_690919.1 | 2, 3 | 159 | ||
| GenBank mRNA | M27883.1 | - | 2, 3 | 159 | ||