siRNA Details
| Gene Symbol: Prrx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153821.1 | NP_722543.1 | 4 | 986 | ||
| XM_006250151.2 | XP_006250213.1 | 5 | 1796 | |||
| GenBank mRNA | AY730764.1 | - | 5 | 782 | ||
| S82911.1 | - | 4 | 986 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prrx1| Gene Symbol: Prrx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153821.1 | NP_722543.1 | 4 | 986 | ||
| XM_006250151.2 | XP_006250213.1 | 5 | 1796 | |||
| GenBank mRNA | AY730764.1 | - | 5 | 782 | ||
| S82911.1 | - | 4 | 986 | |||