siRNA Details
Gene Symbol: Prrx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153821.1 | NP_722543.1 | 1 | 434 | ||
XM_006250151.2 | XP_006250213.1 | 1 | 1172 | |||
GenBank mRNA | AY730764.1 | - | 1 | 158 | ||
S82911.1 | - | 1 | 434 |
