siRNA Details
Gene Symbol: Rims4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170666.1 | NP_733766.1 | 4 | 366 | ||
XM_008762471.1 | XP_008760693.1 | 4 | 483 | |||
GenBank mRNA | AF548739.1 | - | 4 | 366 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rims4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170666.1 | NP_733766.1 | 4 | 366 | ||
XM_008762471.1 | XP_008760693.1 | 4 | 483 | |||
GenBank mRNA | AF548739.1 | - | 4 | 366 |