siRNA Details
Gene Symbol: Kcnj14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170718.1 | NP_733836.1 | 2 | 1454 | ||
GenBank mRNA | AJ003065.1 | - | 2 | 1454 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Kcnj14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170718.1 | NP_733836.1 | 2 | 1454 | ||
GenBank mRNA | AJ003065.1 | - | 2 | 1454 |