siRNA Details
| Gene Symbol: Ppif | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172243.1 | NP_758443.1 | 4 | 387 | ||
| GenBank mRNA | BC086977.1 | - | 4 | 420 | 7193156 | |
| U68544.1 | - | 4 | 387 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppif| Gene Symbol: Ppif | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172243.1 | NP_758443.1 | 4 | 387 | ||
| GenBank mRNA | BC086977.1 | - | 4 | 420 | 7193156 | |
| U68544.1 | - | 4 | 387 | |||