siRNA Details
| Gene Symbol: Sorbs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005762.1 | NP_001005762.1 | 14 | 1316 | ||
| XM_006252241.2 | XP_006252303.1 | 15 | 3000 | |||
| XM_006252242.1 | XP_006252304.1 | 14 | 1407 | |||
| XM_006252244.2 | XP_006252306.1 | 13 | 1635 | |||
| XM_006252246.1 | XP_006252308.1 | 14 | 1352 | |||
| XM_006252248.1 | XP_006252310.1 | 2 | 200 | |||
| XM_008770819.1 | XP_008769041.1 | 12 | 3109 | |||
| GenBank mRNA | AF439378.1 | - | 13 | 1132 | ||
| BC083666.1 | - | 14 | 1316 | 7135547 | ||

Sorbs3
Genomic Map