siRNA Details
| Gene Symbol: Asic3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173135.1 | NP_775158.1 | 7 | 1266 | ||
| XM_008762632.1 | XP_008760854.1 | 7 | 1282 | |||
| GenBank mRNA | AF013598.1 | - | 7 | 1266 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Asic3| Gene Symbol: Asic3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173135.1 | NP_775158.1 | 7 | 1266 | ||
| XM_008762632.1 | XP_008760854.1 | 7 | 1282 | |||
| GenBank mRNA | AF013598.1 | - | 7 | 1266 | ||