siRNA Details
Gene Symbol: Vom1r102 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173298.2 | NP_775420.2 | 1 | 22 | ||
GenBank mRNA | AY510305.1 | - | 1 | 22 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Vom1r102 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173298.2 | NP_775420.2 | 1 | 22 | ||
GenBank mRNA | AY510305.1 | - | 1 | 22 |