siRNA Details
| Gene Symbol: Zfp689 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173330.1 | NP_775452.1 | 3 | 628 | ||
| GenBank mRNA | AF277902.1 | - | 3 | 628 | ||
| BC168646.1 | - | 3 | 636 | 9098465 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp689| Gene Symbol: Zfp689 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173330.1 | NP_775452.1 | 3 | 628 | ||
| GenBank mRNA | AF277902.1 | - | 3 | 628 | ||
| BC168646.1 | - | 3 | 636 | 9098465 | ||