siRNA Details
| Gene Symbol: Nudcd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009621.2 | NP_001009621.1 | 2, 3 | 516 | ||
| GenBank mRNA | BC088299.1 | - | 2, 3 | 516 | 7323100 | |
| FQ216253.1 | - | 2, 3 | 302 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nudcd2| Gene Symbol: Nudcd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009621.2 | NP_001009621.1 | 2, 3 | 516 | ||
| GenBank mRNA | BC088299.1 | - | 2, 3 | 516 | 7323100 | |
| FQ216253.1 | - | 2, 3 | 302 | |||