siRNA Details
Gene Symbol: LOC100910318 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271111.1 | NP_001258040.1 | 2 | 647 | ||
NM_001271112.1 | NP_001258041.1 | 2 | 449 | |||
XM_006246196.2 | XP_006246258.1 | 2 | 408 | |||
XM_006246197.2 | XP_006246259.1 | 2 | 666 |
