siRNA Details
| Gene Symbol: Jmjd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105784.1 | NP_001099254.1 | 5 | 848 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Jmjd4| Gene Symbol: Jmjd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105784.1 | NP_001099254.1 | 5 | 848 | ||