siRNA Details
| Gene Symbol: Nt5m | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105785.1 | NP_001099255.1 | 2 | 264 | ||
| XM_006246460.2 | XP_006246522.1 | 2 | 711 | |||
| GenBank mRNA | BC167058.1 | - | 2 | 320 | 7310205 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nt5m| Gene Symbol: Nt5m | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105785.1 | NP_001099255.1 | 2 | 264 | ||
| XM_006246460.2 | XP_006246522.1 | 2 | 711 | |||
| GenBank mRNA | BC167058.1 | - | 2 | 320 | 7310205 | |