siRNA Details
| Gene Symbol: Mnt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105807.1 | NP_001099277.1 | 3, 4 | 1115 | ||
| XM_008767959.1 | XP_008766181.1 | 3, 4 | 11762 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mnt| Gene Symbol: Mnt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105807.1 | NP_001099277.1 | 3, 4 | 1115 | ||
| XM_008767959.1 | XP_008766181.1 | 3, 4 | 11762 | |||