siRNA Details
| Gene Symbol: Fam101b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007611.1 | NP_001007612.1 | 3 | 558 | ||
| GenBank mRNA | BC079330.1 | - | 3 | 558 | 7096482 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam101b| Gene Symbol: Fam101b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007611.1 | NP_001007612.1 | 3 | 558 | ||
| GenBank mRNA | BC079330.1 | - | 3 | 558 | 7096482 | |