siRNA Details
| Gene Symbol: Pigx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100651.1 | NP_001094121.1 | 3 | 396 | ||
| GenBank mRNA | AB177393.1 | - | 3 | 396 | ||
| DY311520.1 | - | 3 | 394 | |||
| FQ229771.1 | - | 429 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pigx| Gene Symbol: Pigx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100651.1 | NP_001094121.1 | 3 | 396 | ||
| GenBank mRNA | AB177393.1 | - | 3 | 396 | ||
| DY311520.1 | - | 3 | 394 | |||
| FQ229771.1 | - | 429 | ||||