siRNA Details
| Gene Symbol: Hcls1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011898.1 | NP_001011898.1 | 13 | 1304 | ||
| GenBank mRNA | BC079081.1 | - | 13 | 1304 | 7122756 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hcls1| Gene Symbol: Hcls1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011898.1 | NP_001011898.1 | 13 | 1304 | ||
| GenBank mRNA | BC079081.1 | - | 13 | 1304 | 7122756 | |