siRNA Details
Gene Symbol: Hoxd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105885.1 | NP_001099355.1 | 5 | 1479 | ||
XM_008761882.1 | XP_008760104.1 | 2 | 6187 | |||
XM_008761883.1 | XP_008760105.1 | 2 | 4914 | |||
XM_008761884.1 | XP_008760106.1 | 3 | 5666 | |||
XM_008761885.1 | XP_008760107.1 | 3 | 6088 | |||
XM_008761886.1 | XP_008760108.1 | 3 | 4101 | |||
XM_008761887.1 | XP_008760109.1 | 4 | 4002 |
