siRNA Details
| Gene Symbol: Wrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199373.1 | NP_955405.1 | 3 | 364 | ||
| GenBank mRNA | BC062049.1 | - | 3 | 364 | 5598163 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wrb| Gene Symbol: Wrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199373.1 | NP_955405.1 | 3 | 364 | ||
| GenBank mRNA | BC062049.1 | - | 3 | 364 | 5598163 | |