siRNA Details
| Gene Symbol: RGD1306954 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008288.1 | NP_001008289.1 | 6 | 830 | ||
| GenBank mRNA | BC085340.1 | - | 6 | 830 | 7302628 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1306954| Gene Symbol: RGD1306954 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008288.1 | NP_001008289.1 | 6 | 830 | ||
| GenBank mRNA | BC085340.1 | - | 6 | 830 | 7302628 | |