siRNA Details
| Gene Symbol: Srrd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006249579.2 | XP_006249641.1 | 5 | 709 | ||
| XM_213799.8 | XP_213799.6 | 5 | 709 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Srrd| Gene Symbol: Srrd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006249579.2 | XP_006249641.1 | 5 | 709 | ||
| XM_213799.8 | XP_213799.6 | 5 | 709 | |||