siRNA Details
| Gene Symbol: Tmco1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009631.1 | NP_001009631.1 | 259 | |||
| GenBank mRNA | AF531432.1 | - | 182 | |||
| BC088319.1 | - | 259 | 7325542 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmco1| Gene Symbol: Tmco1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009631.1 | NP_001009631.1 | 259 | |||
| GenBank mRNA | AF531432.1 | - | 182 | |||
| BC088319.1 | - | 259 | 7325542 | |||