siRNA Details
| Gene Symbol: Mrps18c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105996.1 | NP_001099466.1 | 1 | 63 | ||
| GenBank mRNA | FQ224447.1 | - | 196 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mrps18c| Gene Symbol: Mrps18c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105996.1 | NP_001099466.1 | 1 | 63 | ||
| GenBank mRNA | FQ224447.1 | - | 196 | |||