siRNA Details
| Gene Symbol: Xbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004210.2 | NP_001004210.1 | 5 | 729 | ||
| NM_001271731.1 | NP_001258660.1 | 6 | 703 | |||
| GenBank mRNA | BC079450.1 | - | 5 | 710 | 7115933 | |
| CB691171.1 | - | 4 | 301 | |||

Xbp1
Genomic Map