siRNA Details
| Gene Symbol: Emc9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008296.1 | NP_001008297.1 | 2 | 401 | ||
| XM_008770673.1 | XP_008768895.1 | 1 | 294 | |||
| GenBank mRNA | BC086432.1 | - | 2 | 401 | 7301648 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Emc9| Gene Symbol: Emc9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008296.1 | NP_001008297.1 | 2 | 401 | ||
| XM_008770673.1 | XP_008768895.1 | 1 | 294 | |||
| GenBank mRNA | BC086432.1 | - | 2 | 401 | 7301648 | |