siRNA Details
| Gene Symbol: Pdlim2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007622.1 | NP_001007623.1 | 3 | 264 | ||
| XM_006252255.2 | XP_006252317.1 | 3 | 282 | |||
| XM_006252256.2 | XP_006252318.1 | 3 | 274 | |||
| XM_006252257.2 | XP_006252319.1 | 3 | 796 | |||
| XM_006252258.1 | XP_006252320.1 | 3 | 164 | |||
| XM_006252259.2 | XP_006252321.1 | 3 | 831 | |||
| XM_008770823.1 | XP_008769045.1 | 3 | 165 | |||
| GenBank mRNA | AY531526.1 | - | 3 | 246 | ||
| BC079091.1 | - | 3 | 264 | 7123600 | ||

Pdlim2
Genomic Map