siRNA Details
Gene Symbol: Thoc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106059.1 | NP_001099529.1 | 4 | 711 | ||
GenBank mRNA | BC168852.1 | - | 4 | 694 | 7443262 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Thoc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106059.1 | NP_001099529.1 | 4 | 711 | ||
GenBank mRNA | BC168852.1 | - | 4 | 694 | 7443262 |