siRNA Details
Gene Symbol: Gins4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030027.1 | NP_001025198.1 | 7 | 663 | ||
XM_006253313.2 | XP_006253375.1 | 7 | 686 | |||
GenBank mRNA | BC099741.1 | - | 7 | 663 | 5623828 |
