siRNA Details
| Gene Symbol: Slc35d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006253493.2 | XP_006253555.1 | 1 | 614 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc35d2| Gene Symbol: Slc35d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006253493.2 | XP_006253555.1 | 1 | 614 | ||