siRNA Details
| Gene Symbol: RGD1307537 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013885.2 | NP_001013907.1 | 3 | 582 | ||
| GenBank mRNA | BC079269.1 | - | 583 | 7133712 | ||
| BF408798.1 | - | 1 | 266 | |||
| CK596086.1 | - | 3 | 582 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1307537| Gene Symbol: RGD1307537 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013885.2 | NP_001013907.1 | 3 | 582 | ||
| GenBank mRNA | BC079269.1 | - | 583 | 7133712 | ||
| BF408798.1 | - | 1 | 266 | |||
| CK596086.1 | - | 3 | 582 | |||