siRNA Details
Gene Symbol: Mzb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024240.1 | NP_001019411.1 | 2 | 194 | ||
GenBank mRNA | BC093397.1 | - | 2 | 194 | 7389084 | |
FQ234743.1 | - | 2 | 228 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mzb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024240.1 | NP_001019411.1 | 2 | 194 | ||
GenBank mRNA | BC093397.1 | - | 2 | 194 | 7389084 | |
FQ234743.1 | - | 2 | 228 |