siRNA Details
| Gene Symbol: Cd63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017125.3 | NP_058821.1 | 4, 5 | 396 | ||
| XM_008765024.1 | XP_008763246.1 | 4, 5 | 458 | |||
| XM_008765025.1 | XP_008763247.1 | 4, 5 | 661 | |||
| GenBank mRNA | AF159104.1 | - | 20 | |||
| BC063173.1 | - | 4, 5 | 399 | 6921594 | ||
| FQ214468.1 | - | 4, 5 | 463 | |||
| FQ221333.1 | - | 4, 5 | 400 | |||
| FQ222038.1 | - | 4, 5 | 379 | |||
| FQ230116.1 | - | 4, 5 | 367 | |||
| X61654.1 | - | 4, 5 | 396 | |||

Cd63
Genomic Map