siRNA Details
| Gene Symbol: Cnep1r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106173.1 | NP_001099643.1 | 2, 3 | 190 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cnep1r1| Gene Symbol: Cnep1r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106173.1 | NP_001099643.1 | 2, 3 | 190 | ||