siRNA Details
| Gene Symbol: Cnep1r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106173.1 | NP_001099643.1 | 3 | 238 | ||
| XM_008772374.1 | XP_008770596.1 | 2 | 173 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cnep1r1| Gene Symbol: Cnep1r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106173.1 | NP_001099643.1 | 3 | 238 | ||
| XM_008772374.1 | XP_008770596.1 | 2 | 173 | |||