siRNA Details
Gene Symbol: Galr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019173.1 | NP_062046.1 | 2 | 319 | ||
XM_006241972.2 | XP_006242034.1 | 1 | 482 | |||
GenBank mRNA | AF031522.1 | - | 1 | 294 | ||
AF073798.1 | - | 2 | 319 | |||
AF079844.1 | - | 1 | 294 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
