siRNA Details
Gene Symbol: Ccdc97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106235.1 | NP_001099705.1 | 5 | 1111 | ||
GenBank mRNA | FQ212311.1 | - | 5 | 1035 | ||
FQ232330.1 | - | 5 | 1031 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ccdc97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106235.1 | NP_001099705.1 | 5 | 1111 | ||
GenBank mRNA | FQ212311.1 | - | 5 | 1035 | ||
FQ232330.1 | - | 5 | 1031 |