siRNA Details
| Gene Symbol: Olr13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000116.1 | NP_001000116.1 | 1 | 324 | ||
| Gene Symbol: LOC100909966 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003748916.2 | XP_003748964.1 | 336 | |||

Olr13
Genomic Map