siRNA Details
| Gene Symbol: Lyve1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106286.1 | NP_001099756.1 | 2 | 527 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lyve1| Gene Symbol: Lyve1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106286.1 | NP_001099756.1 | 2 | 527 | ||