siRNA Details
Gene Symbol: Fam57b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106296.2 | NP_001099766.2 | 1 | 210 | ||
GenBank mRNA | BC161920.1 | - | 1 | 210 | 8370175 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fam57b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106296.2 | NP_001099766.2 | 1 | 210 | ||
GenBank mRNA | BC161920.1 | - | 1 | 210 | 8370175 |