siRNA Details
| Gene Symbol: LOC102554586 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759935.1 | XP_008758157.1 | 2 | 66 | ||
| GenBank mRNA | EX491102.1 | - | 2 | 545 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102554586| Gene Symbol: LOC102554586 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759935.1 | XP_008758157.1 | 2 | 66 | ||
| GenBank mRNA | EX491102.1 | - | 2 | 545 | ||